Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ EDTA, Disodium Salt, Dihydrate, OmniPur™, Calbiochem™,
Click to view available options
Quantity:
250 g
1 kg
Specifications
Melting Point | 252°C |
Color | White |
Odor | Odorless |
Quantity | 250 g |
Solubility Information | Water: Soluble |
Formula Weight | 372.2 |
Percent Purity | ≥99.0% |
Physical Form | Solid |
Chemical Name or Material | EDTA, Disodium Salt, Dihydrate |
Grade | Molecular biology grade |
Safety and Handling
Recommended Storage | Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction