Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA Standard Solution, 1mL = 1mg CaCO3, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Cubetainer
Narrow Mouth Polyethylene Bottle
6381-92-6
C10H18N2Na2O10
372.238
OVBJJZOQPCKUOR-UHFFFAOYSA-L
44120005
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+]
6381-92-6
C10H18N2Na2O10
372.238
OVBJJZOQPCKUOR-UHFFFAOYSA-L
44120005
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 372.24
Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Polyethylene Bottle
Quantity 1L
Specific Gravity 1L = 1.00kg
Chemical Name or Material EDTA Standard Solution, 1mL = 1mg CaCO3