Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EDTA Tetrasodium Salt Dihydrate, 99-101%, Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
1kg
25kg
Packaging:
Plastic bottle
10378-23-1
C10H20N2Na4O10
420.233
MFCD00150026
DGWOWDJDIBPYOR-UHFFFAOYSA-N
ethylenediaminetetraacetic acid tetrasodium salt dihydrate,tetrasodium ethylenediaminetetraacetate dihydrate,edta-na4,edta tetrasodium salt,edta tetrasodium salt hydrate,ethylenediaminetetraacetic acid tetrasodium salt hydrate,64-02-8 anhydrous,ksc909a7h,edta tetrasodium salt dihydrate,tetrapotassium ion 4-edta dihydrate
131843362
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;dihydrate
C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.O.[Na].[Na].[Na].[Na]
10378-23-1
C10H20N2Na4O10
420.233
MFCD00150026
DGWOWDJDIBPYOR-UHFFFAOYSA-N
ethylenediaminetetraacetic acid tetrasodium salt dihydrate,tetrasodium ethylenediaminetetraacetate dihydrate,edta-na4,edta tetrasodium salt,edta tetrasodium salt hydrate,ethylenediaminetetraacetic acid tetrasodium salt hydrate,64-02-8 anhydrous,ksc909a7h,edta tetrasodium salt dihydrate,tetrapotassium ion 4-edta dihydrate
131843362
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;dihydrate
C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.O.[Na].[Na].[Na].[Na]
Formula Weight 416.20
Physical Form Crystalline Powder
Percent Purity 99 to 101%
Packaging Plastic bottle
Quantity 1kg
Chemical Name or Material Ethylenediaminetetraacetic Acid Tetrasodium Salt Dihydrate