Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDTA Titrant, 0.0100 M (M/100), Ricca Chemical

Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
16 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Color | Colorless |
CAS Min % | 98.0, 0.36 |
CAS Max % | 100.0, 0.38 |
Packaging | Natural Poly Bottle |
Quantity | 16 L |
Physical Form | Liquid |
Concentration or Composition Notes | 0.0100Molar |
Chemical Name or Material | EDTA Titrant |