Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA Titrant, 0.0100 Molar, Ricca Chemical

Click to view available options
Quantity:
500mL
1L
4L
10L
12L
16L
20L
Packaging:
Cubitainer
Cubitainer™
Poly natural
6381-92-6,7732-18-5
C10H18N2Na2O10
372.238
OVBJJZOQPCKUOR-UHFFFAOYSA-L
edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
44120005
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+]
6381-92-6,7732-18-5
C10H18N2Na2O10
372.238
OVBJJZOQPCKUOR-UHFFFAOYSA-L
edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
44120005
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+]
CAS Min % 98.0,0.36
CAS Max % 100.0,0.38
Physical Form Liquid
Packaging Poly natural
Concentration or Composition Notes 0.0100Molar
Color Colorless
Quantity 12L
Chemical Name or Material EDTA Titrant