Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA Titrant, 0.0250 M (M/40), Ricca Chemical
SDP

Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
6381-92-6, 7732-18-5
C10H18N2Na2O10
372.24
MFCD00150037,MFCD00003541
OVBJJZOQPCKUOR-UHFFFAOYSA-L
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
6381-92-6, 7732-18-5
C10H18N2Na2O10
372.24
MFCD00150037,MFCD00003541
OVBJJZOQPCKUOR-UHFFFAOYSA-L
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Color Colorless
PubChem CID 131664146
Packaging Cubitainer
Quantity 20 L
Physical Form Liquid
Concentration or Composition Notes 0.0250Molar
Chemical Name or Material EDTA Titrant
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit