Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA, Tocris Bioscience™

Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
60-00-4
C10H16N2O8
292.24
MFCD00003541
KCXVZYZYPLLWCC-UHFFFAOYSA-N
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
6049
CHEBI:42191
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
This item is not returnable. View return policy
60-00-4
C10H16N2O8
292.24
MFCD00003541
KCXVZYZYPLLWCC-UHFFFAOYSA-N
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
6049
CHEBI:42191
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Chelating agent

Chelating agent; sequesters di- and trivalent metal ions.
Quantity 1 g
Formula Weight 292.24
Percent Purity >98%
Chemical Name or Material EDTA

Recommended Storage : Store at Room Temperature