Learn More
DWK Life Sciences Kimble™ Education-Grade Burets
These economical and versatile burets are ideal for use in educational institutions and for many general laboratory procedures.
Manufacturer:  DWK Life Sciences 17040K50
Catalog No. 11-555-250
Add to cart
Description
Description
  • 17040K supplied with a PTFE stopcock plug
  • Easy-to-read durable black enamel scale for 17040K and 17021H-50
  • 17121H supplied with a PTFE threaded stopcock assembly and a precision ground tip
  • 17021H supplied with a PTFE compression fit stopcock assembly and a precision ground tip
  • 17021H, 25 mL buret has a permanent brown stained scale
  • KIM-KAP dust cap is included
  • Funnel fill style buret
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • The stopcock assemblies for the 17121H and 17021H series burets should be wetted before installation.
  • 50mL, 738mm LENGTH, 0.1mL Graduations, 16mm KIM-KAP™, case of 12
Specifications
Specifications
Glass
Education Grade Buret