Learn More
Edvotek™ Dual DNA Electrophoresis LabStation™ Products for Science Education
With the Edvotek Dual DNA Electrophoresis LabStation, seamless, injection-molded horizontal agarose electrophoresis units allow more than one lab group to run gels simultaneously.
Manufacturer:  Edvotek™ 5063
Catalog No. S68646
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Supports up to 16 students
Specifications
Specifications
Two M12 electrophoresis apparatus with one 7 x 14cm gel tray, One Edvotek™ DuoSource™ 150 dual-power source (75/150V, for one or two unit), two variable micropipets, 5 to 50μL
75/150V
Dual DNA electrophoresis LabStation™
400mL
2.7 in.
M12
Lifetime
13.5 oz.
14cm
7cm