Learn More
Edvotek™ Reagents for Electrophoresis: Restriction Enzyme Reaction Buffer
For restriction enzymes.
Supplier: Edvotek™ 610
Includes: 2 ml 10x Restriction Enzyme Reaction Buffer; Sufficient for 200 Reactions
Description
This reaction buffer is a 10-fold concentrate which contains tris-HCl, pH 8.0, sodium chloride, magnesium chloride and protein stabilizers. The buffer has been optimized for use with Eco RI, BAM HI, and Hind III restriction enzymes.Specifications
10-fold concentrate which contains tris-HCl, pH 8.0, sodium chloride, magnesium chloride and protein stabilizers | |
Can be stored at room temperature. | |
200 Reactions | |
Restriction Enzyme Reaction Buffer |