Learn More
Edwards Fittings for BOC High Vacuum Pumps: Blank Flange
For use with BOC high vacuum pumps
Manufacturer:  Edwards C10512368
 View more versions of this product
Catalog No. 11-283-16
Add to cart
Description
Description
  • Made of aluminum
  • Aluminum is ideal for achieving dependable cost-effective performance down to 10-7mbar
Specifications
Specifications
BOC high vacuum pump
Aluminum
30 x 17.2 x 6 x 2.5 mm
NW16 Blank Flange