Learn More
Edwards Fittings for BOC High Vacuum Pumps: Centering Ring
For use with BOC high vacuum pumps
Supplier: Edwards C10516395
Description
- Aluminum centering rings manufactured from high-tech polymer
- Unique slotted design prevents gas bursts
- CX2 polymer can be used at temperatures up to 120°C and is unaffected by most common solvents
- Outgassing rate of the material is 6.6 × 10-8mbar l s-1cm-2
Specifications
BOC high vacuum pump | |
Stainless Steel | |
NW40 Centering Ring |