Learn More
Edwards Fittings for BOC High Vacuum Pumps: Short Flange
For use with BOC high vacuum pumps
Supplier: Edwards C10512311
Description
- Made of aluminum
- Aluminum is ideal for achieving dependable cost-effective performance down to 10-7mbar
Specifications
BOC high vacuum pump | |
Stainless Steel | |
NW16 Short Flange |