Learn More
Edwards Fittings for BOC High Vacuum Pumps: Swing Clamp
For use with BOC high vacuum pumps
Manufacturer:  Edwards C10517304
 View more versions of this product
Catalog No. 11-283-36
Add to cart
Description
Description
  • Compact stainless-steel clamping ring
  • Lightweight polymer clamp
  • Rugged pneurop standard aluminium clamp
  • Metal clamp suitable for aluminium and indium seals wide temperature range
  • Claw clamps and bolts provide wide range of applications
Specifications
Specifications
BOC High Vacuum Pump
Polymer
120 x 86 x 35 mm
NW50 Swing Clamp