Learn More
Edwards High Vacuum Right Angle Valve
Supplier: Edwards C31205000
Specifications
Body: HE30TF grade Aluminum; Bellows: AISI 316L stainless steel; O ring: fluoroelastomer | |
100°C | |
Right Angle Valve |