Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edwards Rotary Vane Vacuum General-Purpose Pumps: RV3

High-performance pump suitable for a wide range of difficult applications

Manufacturer:  Edwards R14000001

Catalog No. 11-283-79

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Includes: Pump, 2m electrical supply cable, inlet screen and Ultragrade 19 Oil, lifting bracket

Description

Description

Unique dual mode allows you to configure pump for very high gas loads without compromising vacuum. Easily reconfigured—even when pump is running. For centrifugal concentrators, rotary evaporators, freeze dryers, gel dryers, vacuum ovens, or general-purpose requirements.

  • Large, clearly marked, easily accessible controls
  • Operate at quiet 48dBA
  • Easy to use and service (require no special tools)
  • Fast-acting inlet valve prevents oil and gas suckback in the event of power or oil pressure loss
  • NW25 inlet/outlet connections
High vacuum or high throughput from the same pump
  • Easily selectable modes
  • High Vacuum mode provides ultimate vacuum down to 1.5 x 10-3 torr with no gas ballast
  • High Throughput mode sustains long-term high inlet pressures with ultimate vacuum as low as 2.3 x 10-2 torr without gas ballast
Adjustable gas ballast control maximizes vapor handling capability
  • Gas ballast delivered into pump prevents condensation of vapor carried by pumped gases
  • Three user-selectable gas ballast positions—'0' Closed, 'I' Low Flow, 'II' High Flow—meet most vapor handling requirements

Order Info

For 115V 50/60Hz. Vacuum tubing sold separately.

Specifications

Specifications

1.5 x 10-3mbar
115 V
115 V 50/60 Hz
48 dB (A)
21.6 kg
40°C
76 L/min.
9 in.
17 in.
6.25 in.
0.5 hp
Pump, 2m electrical supply cable, inlet screen and Ultragrade 19 Oil, lifting bracket
RV3
47.61 lb.
104°F
12°C
50/60 Hz
22.9 cm
43.18 cm
IP44
Documents
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit