Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EEA1 Mouse anti-Human, Mouse, Alexa Fluor™ 594, Clone: 6D4.1D4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP236568AF594

Catalog No. NP23668A94


Add to cart

Description

Description

EEA1 Monoclonal antibody specifically detects EEA1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry.
Specifications

Specifications

EEA1
6D4.1D4
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular)
Conjugated, Purified
EEA1
Partial recombinant human EEA1 protein (between amino acids 50-300). [UniProt Q15075]
Protein G purified
Autophagy, Cancer, Cardiovascular Biology, Cellular Signaling, Cytokine Research, Endosome Markers, Immunology, Innate Immunity, Lysosome Markers, Membrane Trafficking and Chaperones
Primary
8411.0
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
early endosome antigen 1, early endosome antigen 1, 162kD, early endosome-associated protein, Endosome-associated protein p162, MSTP105, ZFYVE2, ZFYVE2MST105, Zinc finger FYVE domain-containing protein 2
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit