Learn More
EEA1 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
SDP
Rabbit Polyclonal Antibody
Supplier:  Bethyl Laboratories, Inc A301897A
Encompass_Preferred
Catalog No. 50-156-1261
Add to Cart
Description
Description
The recommended shelf life for this product is 1 year from date of receipt.
Binds phospholipid vesicles containing phosphatidylinositol 3-phosphate and participates in endosomal trafficking.
Specifications
Specifications
EEA1
Polyclonal
Unconjugated
EEA1
EEA1, Endosome-associated protein p162, Zinc finger FYVE domain-containing protein 2
Rabbit
Antigen affinity chromatography
RUO
216238, 8411
4° C
Liquid
Immunoprecipitation, Western Blot
0.2 mg/ml
TBS with 0.1% BSA and 0.09% sodium azide
Q15075, Q8BL66
Eea1
Between 175 and 225
100 μL
Primary
Human, Mouse
Antibody
IgG