Learn More
EEA1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 222661AP
Catalog No. 50-173-3805
Add to cart
Description
Description
Early endosomal antigen 1 (EEA1) is a 162 kDa membrane bound protein component specific to the early endosome and is essential for fusion between early endocytic vesicles. Early endosomes are cytoplasmic compartments which fuse with endocytic vesicles for redistribution of extracellular compounds to alternate destinations. Zinc-finger-like domains, reminiscent of those found in nucleic acid binding proteins, are located in the amino and carboxyl-terminal domains of EEA1. The carboxyl-terminal zinc-finger-like-domain is conserved in several other non-nuclear proteins, some of which are also involved in intracellular protein trafficking. In addition, this domain is an authentic zinc-binding domain and is critical to the intracellular localization of EEA1. Membrane association of EEA1 has been shown dependent on phosphatidyl 3-kinase activity, inhibitors of which cause EEA1 to dissociate from early endosomes.
Specifications
Specifications
EEA1
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q15075, Q8BL66
early endosome antigen 1, EEA1, MST105, MSTP105, ZFYVE2
Rabbit
IgG
150 μL
-20°C
Primary
216238, 8411
ELISA
Unconjugated
EEA1
Liquid
EEA1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse