Learn More
EEA1 Rabbit anti-Human, Rat, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29947
Catalog No. 89-033-235
Add to cart
Description
Description
Rabbit polyclonal antibody raised against synthetic peptide of human EEA1.
  • High specificity
  • High reactivity
Specifications
Specifications
EEA1
Unconjugated
Immunofluorescence (1:100) Immunoprecipitation (1:500) Western Blot (0.06 ug/mL) The optimal working dilution should be determined by the end user.
NM_003566
MST105|MSTP105|ZFYVE2
Rabbit
Affinity purification
RUO
Primary
8411
Immunofluorescence, Immunoprecipitation, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of human EEA1.
In PBS (2% sucrose, 0.09% sodium azide).
Liquid
EEA1
A synthetic peptide corresponding to N-terminus of human EEA1.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Rat
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)