Learn More
EEF1B2 Rabbit anti-Human, FITC, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP300341F
Catalog No. NB065640
Add to cart
Description
Description
EEF1B2 Polyclonal specifically detects EEF1B2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
EEF1B2
FITC
PBS
EEF1B, EEF1B1, EF1B, EF-1-beta, elongation factor 1-beta, eukaryotic translation elongation factor 1 beta 1, eukaryotic translation elongation factor 1 beta 2
Produced in rabbits immunized with purified, recombinant EEF1B2 (P24534; Met1-Ile225).
Affinity purified
RUO
Primary
1933
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human