Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ EEF1G Recombinant Protein
$503.00 - $775.00
Specifications
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
---|---|
Molecular Weight (g/mol) | 76.2 |
pH Range | 8 |
Preparation Method | in vitro wheat germ expression system |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-010-050
|
Abnova™
H00001937P0110 |
10 ug |
Each for $503.00
|
|
|||||
89-010-051
|
Abnova™
H00001937P0125 |
25μg |
Each for $775.00
|
|
|||||
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Human | |
Solution |
76.2 | |
in vitro wheat germ expression system | |
Wheat Germ (in vitro) | |
Recombinant |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title