Learn More
EEF2 Mouse anti-Human, Unconjugated, Clone: 1E4D5, Proteintech
Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 675501IG150UL
This item is not returnable.
View return policy
Description
The EEF2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EEF2. This antibody recognizes human, mouse, rat antigen. The EEF2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
EEF2 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
1E4D5 | |
Unconjugated | |
150 μL | |
1938 | |
IgG1 |