Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ EEF2K Human Recombinant Protein, TMW: 84.6kDa

Highly purified. Generating reliable and reproducible results.

$344.00 - $976.00

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-Page
Formulation Phosphate buffered saline (pH7.4), 10% glycerol
Gene ID (Entrez) 29904
Molecular Weight TMW: 84.6kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25848002
View Documents
Novus Biologicals™
NBP2518480.02MG
0.02mg
Each for $344.00
Add to cart
 
NBP25184801
View Documents
Novus Biologicals™
NBP2518480.1MG
0.1mg
Each for $976.00
Add to cart
 
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-725 of Human EEF2K The Recombinant Human EEF2K Protein is derived from E. coli. The Recombinant Human EEF2K Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

0.25mg/mL
Phosphate buffered saline (pH7.4), 10% glycerol
TMW: 84.6kDa
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
EEF2K
Yes
SDS-Page
29904
>85%, by SDS-PAGE
Calcium/calmodulin-dependent eukaryotic elongation factor 2 kinase, calcium/calmodulin-dependent eukaryotic elongation factor-2 kinase, EC 2.7.11, EC 2.7.11.20, eEF-2 kinase, eEF-2Kcalmodulin-dependent protein kinase III, elongation factor-2 kinase, eukaroytic elongation factor 2 kinase, eukaryotic elongation factor 2 kinase, eukaryotic elongation factor-2 kinase, HSU93850, MGC45041
Unlabeled
SDS
Product Certifications

For research use only and is not approved for use in humans or in clinical diagnosis

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit