Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EEN Antibody (GT3065), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313631
This item is not returnable.
View return policy
Description
EEN Monoclonal antibody specifically detects EEN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| EEN | |
| Monoclonal | |
| Unconjugated | |
| CNSA1endophilin-2, EEN fusion partner of MLL, EENSH3 domain-containing GRB2-like protein 1, Endophilin-2, extra 11-19 leukemia fusion, Extra eleven-nineteen leukemia fusion gene protein, MGC111371, SH3 domain protein 2B, SH3-containing Grb-2-like 1 protein, SH3D2Bendophilin-A2, SH3-domain GRB2-like 1, SH3P8 | |
| Recombinant protein encompassing a sequence within the center region of mouse EEN. The exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| GT3065 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 6455 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction