Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Tocris Bioscience™ Efaroxan hydrochloride Available on GSA/VA Contract for Federal Government customers only.
α2 antagonist and I1, I3 ligand
Manufacturer:  Tocris Bioscience™ 0792/10
 View more versions of this product
Catalog No. 07-921-0
Description
Description
Potent, highly selectiveα2 adrenoceptor antagonist and imidazoline I1 receptor ligand (pKi values are 7.87, 7.42, 5.74, 7.28 and < 5 forα 2A,α2B,α2C, I1, and I2 receptors respectively). Promotes insulin secretion, at a site distinct from I1 or I2 (the putative I3 receptor) in vitro and in vivo.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
2-(2-Ethyl-2,3-dihydro-2-benzofuranyl)-4,5-dihydro-1H-imidazole hydrochloride
>99%
Observed MW: 252.74
Soluble to 100 mM in water
89197-00-2
C13H16N2O.HCl
10mg
Safety and Handling
Safety and Handling
Recommended Storage : Store at RT