Learn More
Tocris Bioscience™ Efaroxan hydrochloride Available on GSA/VA Contract for Federal Government customers only.
$142.80 - $600.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
07-925-0 View Documents Tocris Bioscience™
0792/50
Each for $600.00
Add to cart
07-921-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
0792/10
Each for $142.80
Add to cart
Description
Description
Potent, highly selective α2 adrenoceptor antagonist and imidazoline I1 receptor ligand (pKi values are 7.87, 7.42, 5.74, 7.28 and < 5 for α 2A, α2B, α2C, I1, and I2 receptors respectively). Promotes insulin secretion, at a site distinct from I1 or I2 (the putative I3 receptor) in vitro and in vivo.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.