Learn More
Tocris Bioscience™ Efaroxan hydrochloride
α2 antagonist and I1, I3 ligand
Manufacturer:  Tocris Bioscience™ 0792/50
 View more versions of this product
Catalog No. 07-925-0
Add to cart
Description
Description
Potent, highly selectiveα2 adrenoceptor antagonist and imidazoline I1 receptor ligand (pKi values are 7.87, 7.42, 5.74, 7.28 and < 5 forα 2A,α2B,α2C, I1, and I2 receptors respectively). Promotes insulin secretion, at a site distinct from I1 or I2 (the putative I3 receptor) in vitro and in vivo.
Specifications
Specifications
2-(2-Ethyl-2,3-dihydro-2-benzofuranyl)-4,5-dihydro-1H-imidazole hydrochloride
>99%
Observed MW: 252.74
Soluble to 100 mM in water
89197-00-2
C13H16N2O.HCl
50mg
Safety and Handling
Safety and Handling
Recommended Storage : Store at RT