Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFCAB7 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548662

Catalog No. PA548662

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

EFCAB7
Polyclonal
PBS with 0.09% sodium azide
A8K855, Q8VDY4
EF-hand calcium-binding domain-containing protein 7, EFCAB7, KIAA1799
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
230500, 84455
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
EFCAB7
Liquid
EFCAB7
KLH conjugated synthetic peptide between 434-462 amino acids from the C-terminal region of human EFCB7.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit