Learn More
EFD™ Ultra Dispenser - Precision Tips
Burr-free, polished and passivated stainless steel tips with polypropylene SafetyLok™ hubs
Manufacturer:  EFD 5122B
 View more versions of this product
Catalog No. 13-850-109
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
45°
Precision Tip
22 ga
Stainless Steel
Blue
Ultra dispenser
0.50 in.
12.7mm