Sino Biological™ EFNA3 (EphrinA3) Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding mature form of human EphrinA3, residues 1 to 213, fused topolyhistidine tag at C-terminus

Manufacturer: Sino Biological™ 10188H08H25

 View more versions of this product

Catalog No. 10-188-H08H25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 23kd
    Purity ≥90% by SDS-PAGE
    Quantity 5 x 5μg
    Source Human
    Storage Requirements −20° to −70°C
    Structural Form Recombinant
    Endotoxin Concentration <1 EU/μg
    Common Name Ephrin-A3
    Gene Symbol EFNA3
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Kinases and Inhibitors
    Expression System Mammalian
    Product Type EFNA3 (EphrinA3) Recombinant Human Protein
    For Use With (Application) Studying protein-protein interactions and protein structure analysis
    Format Lyophilized
    Gene ID (Entrez) 1944

    Expressed from DNA sequence encoding mature form of human EphrinA3, residues 1 to 213, fused topolyhistidine tag at C-terminus

    • Activity is measured by its ability to compete with human EFNA3-Fc for binding to immobilized mouse EFNA6-His in functional ELISA assay
    • Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit