Tocris™ Efonidipine hydrochloride monoethanolate

Ca2+ channel blocker (L- and T-type)

Manufacturer: Tocris™ 3733/50

 View more versions of this product

Catalog No. 37-335-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors Efonidipine hydrochloride monoethanolate
    Purity >99
    Molecular Weight 714.18
    Quantity 50mg
    Product Type Efonidipine hydrochloride monoethanolate

    Selective blocker of L-type and T-type Ca2+ channels. Displays minimal inhibition of N- and P/Q-type channels and no inhibition of R-type channels. R(-) and S(+)-enantiomers display different channel selectivity; S(+)-Efonidipine blocks L-type and T-type channels whereas R(-)-Efonidipine displays selectivity for T-type channels. Exhibits antihypertensive activity.

    • Chemical Name: 5-(5,5-Dimethyl-2-oxido-1,3,2-dioxaphosphorinan-2-yl)-1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3-pyridinecarboxylic acid 2-[phenyl(phenylmethyl)amino]ethyl ester hydrochloride monoethanolate
    • Formula: C34H38N3O7P.HCl.C2H5OH
    • CAS Number: 111011-76-8
    • Solubility: Soluble to 100mM in DMSO
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit