Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EFR3A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554694
Description
Immunogen sequence: LLMVTSGYKA KTIVTALPGS FLDPLLSPSL MEDYELRQLV LEVMHNLMDR HDNRAKLRGI RIIPDVADLK IKREKICRQD TSFMK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Specifications
EFR3A | |
Polyclonal | |
Unconjugated | |
EFR3A | |
A130089M23Rik; BB071175; C76891; C920006C10Rik; D030063F01Rik; EFR3 homolog A; EFR3 homolog A (S. cerevisiae); Efr3a; KIAA0143; mKIAA0143; Protein EFR3 homolog A; Protein EFR3-like; RGD1305976 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23167, 362923, 76740 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14156, Q8BG67 | |
EFR3A | |
Recombinant protein corresponding to Human EFR3A. Recombinant protein control fragment (Product #RP-93789). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction