Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EFR3B Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | EFR3B |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB126441
|
Novus Biologicals
NBP310770100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
EFR3B Polyclonal specifically detects EFR3B in Human samples. It is validated for Western Blot.Specifications
EFR3B | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
EFR3 homolog B (S. cerevisiae), KIAA0953 | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Human EFR3B (NP_055786). Peptide sequence VPSLLFNLQHVEEAESRSPSPLQAPEKEKESPAELAERCLRELLGRAAFG | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
22979 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title