Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFS Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549537

Catalog No. PA549537

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Docking protein which plays a central coordinating role for tyrosine-kinase-based signaling related to cell adhesion. May serve as an activator of SRC and a downstream effector. Interacts with the SH3 domain of FYN and with CRK, SRC, and YES.
Specifications

Specifications

EFS
Polyclonal
PBS with 0.09% sodium azide
O43281
Embryonal Fyn-associated substrate, hEFS, Cas scaffolding protein family member 3, EFS, CASS3
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10278
Western Blot
Unconjugated
EFS
Liquid
EFS
KLH conjugated synthetic peptide between 367-399 amino acids from the Central region of human EFS.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit