Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFTUD1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP276363

Catalog No. NBP276363


Add to cart

Description

Description

EFTUD1 Polyclonal antibody specifically detects EFTUD1 in Human samples. It is validated for Immunoprecipitation.
Specifications

Specifications

EFTUD1
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
EFL1, Elongation Factor Tu GTP Binding Domain Containing 1, Elongation Factor Tu GTP-Binding Domain-Containing Protein 1, Elongation Factor-Like 1, FAM42A, HsT19294, Protein FAM42A, RIA1, Ribosome Assembly 1 Homolog, Ribosome Assembly 1 Homolog (Yeast)
Rabbit
IgG
100 μL
Primary
79631.0
Immunoprecipitation
Immunoprecipitation 15-20 μL/mg lysate
Purified
EFTUD1
Immunogen sequence between amino acids 50 and 100
Antigen Affinity-purified
Store at 2-8°C/ 1 year from date of receipt
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit