Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EG-VEGF/PK1 Antibody (MM0043-2L13) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
$204.00 - $542.50
Specifications
Antigen | EG-VEGF/PK1 |
---|---|
Clone | MM0043-2L13 |
Concentration | LYOPH |
Applications | Immunohistochemistry (Paraffin), Western Blot, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018048
|
Novus Biologicals
NB110935140.025MG |
0.025 mg |
Each for $204.00
|
|
|||||
NB11093514
|
Novus Biologicals
NB11093514 |
0.1 mg |
Each for $542.50
|
|
|||||
Description
EG-VEGF/PK1 Monoclonal specifically detects EG-VEGF/PK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EG-VEGF/PK1 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Mouse, Human, Rat | |
84432 | |
human recombinant EG-VEGF protein | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
MM0043-2L13 | |
Immunohistochemistry (Paraffin), Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication | |
EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, prokineticin 1 | |
PROK1 | |
IgG2 | |
Protein G purified | |
This recognizes human EG-VEGF in western blot. It is highly cross reactive rat and mouse EG-VEGF proteins. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title