Learn More
EG-VEGF/PK1 Mouse anti-Human, Mouse, Rat, DyLight 755, Clone: MM0043-2L13, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11093514IR
Catalog No. NBT93514IR
Add to cart
Description
Description
EG-VEGF/PK1 Monoclonal antibody specifically detects EG-VEGF/PK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
EG-VEGF/PK1
MM0043-2L13
Western Blot, ELISA
Purified
PROK1
human recombinant EG-VEGF protein
Protein G purified
Cell Cycle and Replication
Primary
84432
Human, Mouse, Rat
Western Blot, ELISA
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, prokineticin 1
Mouse
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
This recognizes human EG-VEGF in western blot. It is highly cross reactive rat and mouse EG-VEGF proteins.