Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGF Mouse anti-Human, Unconjugated, Clone: 7C9.14.6, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen EGF
Applications Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF22941MI
View Documents
MilliporeSigma
MABF2294100UG
100 μg
Each for $372.00
Add to cart
 
MABF22942MI
View Documents
MilliporeSigma
MABF229425UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

EGF Monoclonal antibody specifically detects EGF in Human samples. It is validated for Immunohistochemistry (Paraffin), Affinity Binding Assay, Western Blotting.
Specifications

Specifications

EGF
Unconjugated
Mouse
Urogastrone
A recombinant fragment corresponding to 53 amino acids from the C-terminal region of human Epidermal growth factor.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunohistochemistry (Paraffin), Western Blot
Purified
Human
EGF
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit