Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGF Rat anti-Mouse, HRP, Clone: RM0078-2M13, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP121708H
Description
EGF Monoclonal specifically detects EGF in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EGF | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
EGF | |
Mouse recombinant EGF | |
0.1 mL | |
Primary | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant EGF. | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0078-2M13 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG | |
Rat | |
Protein A or G purified | |
Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Virology Bacteria and Parasites | |
1950 | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title