Learn More
EGF Rat anti-Mouse, Janelia Fluor 646, Clone: RM0078-2M13, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP121708JF646
Catalog No. NBP12708646
Add to cart
Description
Description
EGF Monoclonal antibody specifically detects EGF in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
EGF
RM0078-2M13
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
EGF
Mouse recombinant EGF
Protein G purified
Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Virology Bacteria and Parasites
Primary
1950
Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), HOMG4, pro-epidermal growth factor, URG
Rat
IgG2
0.1mL
Store at 4°C in the dark.
Monoclonal
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant EGF.