Learn More
EGFL7 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 192911AP
Catalog No. 50-173-2382
Add to cart
Description
Description
Epidermal growth factor-like protein 7 (EGFL7), also known as VE-statin, MEGF7, Notch4-like protein or Zneu1, is a 29.6 kDa protein produced and secreted by endothelium. The protein is also expressed by some normal and maligt tissues. The main function of EGFL7 is to regulate angiogenesis and vasculogenesis. A high level of EFGL-7 can be found in tissues with active angiogenesis of a variety of disorders.
Specifications
Specifications
EGFL7
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UHF1
EGF like domain, multiple 7, EGF like protein 7, EGFL7, MEGF7, NOTCH4 like protein, RP11 251M1.2, Vascular endothelial statin, VE STATIN, ZNEU1
Rabbit
IgG
150 μL
-20°C
Primary
51162
Immunohistochemistry (Paraffin)
Unconjugated
EGFL7
Liquid
EGFL7
EGFL7 Fusion Protein Ag6559
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human