Learn More
EGFR (ErbB1) [T790M] [L858R] Recombinant Human Protein
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
PV4879 Non-distribution product as customer accommodation.
View Documents
LIFE TECHNOLOGIES
PV4879
N/A 10μg N/A
Description
Description
The epidermal growth factor receptor is a receptor tyrosine kinases that signals in response to various growth factors. Overexpression has been linked to numerous types of cancer and EGFR is the target of both biological and small molecular therapeutics. The double T790M L858R mutant of EGFR displays resistance to Erlotinib (Tarceva) and sensitivity to HKI-272 and rapamycin combination therapy.