Learn More
EGFR Human anti-Human, Alexa Fluor 405, Clone: Hu1, R&D Systems™
Human Monoclonal Antibody
Supplier:  R&D Systems FAB9577V100UG
Catalog No. FAB9577V100
Add to Cart
This item is not returnable. View return policy
Description
Description
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
EGFR
Monoclonal
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Human
Protein A or G purified from cell culture supernatant
Primary
Detects human EGFR based on Cetuximab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Cetuximab. This product is for research use only.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Flow Cytometry
Hu1
Flow Cytometry 0.25-1 ug/10^6 cells
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, EGF R, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ErbB, ErbB1, ERBB1PIG61, HER1, HER-1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1
Human EGF R/ErbB1
100 μg
1956
Human
Supernatant