Learn More
EGFR Human anti-Human, Alexa Fluor 647, Clone: Hu1, R&D Systems™
Human Monoclonal Antibody
Manufacturer:  R&D Systems FAB9577R100
Catalog No. FAB9577R100
Add to cart
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
EGFR
Hu1
Flow Cytometry 5 uL/10^6 cells
Supernatant
Human
IgG1
100 Tests
Primary
1956
Human
Flow Cytometry
Alexa Fluor 647
Supplied in a saline solution containing BSA and Sodium Azide.
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, EGF R, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ErbB, ErbB1, ERBB1PIG61, HER1, HER-1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1
Human EGF R/ErbB1
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human EGFR based on Cetuximab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Cetuximab. This product is for research use only.