Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGFR Human anti-Human, Clone: Matuzumab, Novus Biologicals™

Human Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275911SS

Catalog No. NBP275911SS


Add to cart

Description

Description

EGFR Monoclonal antibody specifically detects EGFR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Block/Neutralize.
Specifications

Specifications

EGFR
Matuzumab
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Block/Neutralize
Purified
EGFR
Domain III of the extracellular domain of human EGRF.
Protein A purified
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Monoclonal
Binds to human EGFR.
Blocking Assay, ELISA, Flow Cytometry, Immunohistochemistry, Neutralization, Western Blot
Unconjugated
PBS with 0.02% Proclin 300
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, biosimilar, biosimilars, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor
Human
IgG1 Kappa
0.025 mg
Primary
1956.0
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit