Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGFR Mouse anti-Human, Alexa Fluor™ 350, Clone: GFR/1708, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP254330AF350
Description
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Western Blot.Specifications
EGFR | |
GFR/1708 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
Mouse | |
IgG1 Kappa | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
EGFR | |
Recombinant human full-length EGFR protein | |
0.1 mL | |
Primary | |
1956 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title