Learn More
EGFR Mouse anti-Human, Clone: SPM622, Abnova™
Mouse monoclonal antibody raised against native human EGFR.
Manufacturer:  Abnova Corporation MAB13062
Catalog No. 89-934-071
Add to cart
Description
Description
The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily. This protein is a receptor for members of the epidermal growth factor family. EGFR is a cell surface protein that binds to epidermal growth factor. Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation. Mutations in this gene are associated with lung cancer. [provided by RefSeq]
Specifications
Specifications
EGFR
SPM622
Mouse monoclonal antibody raised against native human EGFR.
In 10mM PBS (0.05% BSA, 0.05% sodium azide).
ERBB/ERBB1/HER1/PIG61/mENA
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
EGFR
Native purified EGFR from human A431 cells.
Protein A/G purification
RUO
Primary
1956