Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGFR Mouse anti-Human, FITC, Clone: 5E10D3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237326F

Catalog No. NBP237326F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

EGFR
5E10D3
FITC
PBS with 0.05% Sodium Azide
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1
Mouse
IgG2b
Protein A or G purified
Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Hypoxia, Signal Transduction, Tumor Suppressors, Tyrosine Kinases
Primary
1956
ELISA, Flow Cytometry
1 mg/mL
Flow Cytometry 1:10 - 1:1000, ELISA
Purified
EGFR
Purified recombinant fragment of human EGFR (AA: 693-893) expressed in E. Coli.
175 kDa
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit