Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGFR Mouse anti-Human, Unconjugated, Clone: F4, Abnova
Mouse Monoclonal Antibody.
Manufacturer: Abnova Corporation MAB21178
Description
EGFR Monoclonal antibody specifically detects EGFR in Human samples. It is validated for WB.Specifications
EGFR | |
F4 | |
epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian) | |
Liquid | |
Mouse | |
IgG1, Kappa | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. | |
Monoclonal | |
Human |
Western Blot | |
Unconjugated | |
EGFR | |
ERBB, ERBB1, HER1, PIG61, mENA | |
A synthetic peptide corresponding to amino acids 985-996 of human EGFR | |
100 μg | |
Primary | |
1956 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title